Search
Search

Grd| Gene Symbol: Grd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079407.3 | NP_524131.1 | 3 - 4 | 1380 | 67 | |
| GenBank mRNA | BT126369.1 | - | 3 - 4 | 1380 | 67 | |
| X78349.1 | - | 818 | 67 | |||
Set Membership: |
|