Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ?
Create Account
Products
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Tubes
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Clinical Genomics
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO 和 CRO 服務
CDMO 服務
臨床試驗 CRO 服務
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
Register for an Account
How to Order
Instrument Support
Technical Support Centers
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
快速訂購
登入
登入
Don't have an account ?
Create Account
帳戶
訂單
訂製產品與專案
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01823188_m1
See other GIG GE Assays ›
Gene Symbol:
gig
Gene Name:
gigas
Gene Aliases:
6975, C1, CG6975, Dmel\CG6975, Dmel_CG6975, FBpp0074588, Gigas, ME 109, TSC2, Tsc2, Tsc2/gig, dTSC2, dTsc2, gig/Tsc2, gig|Tsc2, l(3)109, tsc2
Chromosome Location:
Chr.3L: 20129181 - 20139010 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0005198
Assay ID
Dm01823188_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 250 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Inventoried
Catalog #
4331182
Price
284.00
Your Price
Online offer:
266.65
266.65
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: gig
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001275160.1
NP_001262089.1
5 - 6
882
84
NM_079453.3
NP_524177.1
5 - 6
882
84
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Recommended Assay
Drosophila melanogaster
Inventoried
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
signal transduction
response to nutrient
negative regulation of cell proliferation
determination of adult lifespan
positive regulation of autophagy
negative regulation of Wnt signaling pathway
negative regulation of cell growth
negative regulation of vascular endothelial growth factor receptor signaling pathway
negative regulation of TOR signaling
larval midgut histolysis
regulation of phosphorylation
cholesterol homeostasis
positive regulation of GTPase activity
negative regulation of cell size
negative regulation of synaptic growth at neuromuscular junction
negative regulation of organ growth
negative regulation of insulin receptor signaling pathway
oogenesis
lymph gland development
regulation of small GTPase mediated signal transduction
regulation of cell cycle
negative regulation of protein kinase B signaling
lipid homeostasis
cellular response to hypoxia
cellular response to chloroquine
negative regulation of TORC1 signaling
positive regulation of lipophagy
positive regulation of TORC2 signaling
regulation of terminal button organization
GTPase activator activity
protein binding
kinase binding
protein kinase binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline