Search
Search

alpha-Cat| Gene Symbol: alpha-Cat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300225.1 | NP_001287154.1 | 4 - 5 | 2300 | 76 | |
| NM_079495.3 | NP_524219.1 | 5 - 6 | 1813 | 76 | ||
| GenBank mRNA | D13964.2 | - | 5 - 6 | 1776 | 76 | |
Set Membership: |
|