Search
Search

stj| Gene Symbol: stj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259374.2 | NP_001246303.1 | 6 - 7 | 1843 | 62 | |
| NM_001259375.2 | NP_001246304.1 | 6 - 7 | 2234 | 62 | ||
| NM_137058.4 | NP_610902.1 | 6 - 7 | 2234 | 62 | ||
| GenBank mRNA | AY069830.1 | - | 6 - 7 | 2554 | 62 | |
Set Membership: |
|