Search
Search

conv| Gene Symbol: conv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299487.1 | NP_001286416.1 | 3 - 4 | 630 | 72 | |
| NM_137104.3 | NP_610948.2 | 3 - 4 | 871 | 72 | ||
| GenBank mRNA | EU814872.1 | - | 3 - 4 | 430 | 72 | |
Set Membership: |
|