Search
Search

cib| Gene Symbol: cib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258587.1 | NP_001245516.1 | 3 - 4 | 674 | 65 | |
| NM_001297923.1 | NP_001284852.1 | 4 - 5 | 428 | 65 | ||
| NM_080326.4 | NP_525065.1 | 3 - 4 | 345 | 65 | ||
| NM_167001.3 | NP_726908.1 | 3 - 4 | 413 | 65 | ||
| GenBank mRNA | AY089633.1 | - | 3 - 4 | 411 | 65 | |
| KX531317.1 | - | 2 - 3 | 252 | 65 | ||
Set Membership: |
|