Search
Search

norpA| Gene Symbol: norpA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014720.2 | NP_001014720.1 | 3 - 4 | 1163 | 89 | |
| NM_001014721.3 | NP_001014721.1 | 2 - 3 | 701 | 89 | ||
| NM_001169190.2 | NP_001162661.1 | 4 - 5 | 1432 | 89 | ||
| NM_001297931.1 | NP_001284860.1 | 3 - 4 | 826 | 89 | ||
| NM_080330.4 | NP_525069.2 | 2 - 3 | 701 | 89 | ||
| NM_167008.2 | NP_726925.1 | 3 - 4 | 1163 | 89 | ||
| GenBank mRNA | AF181641.1 | - | 2 - 3 | 684 | 89 | |
Set Membership: |
|