Search
Search

spirit| Gene Symbol: spirit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169236.2 | NP_001162707.1 | 3 - 4 | 463 | 63 | |
| NM_001272413.1 | NP_001259342.1 | 3 - 4 | 512 | 63 | ||
| NM_132264.3 | NP_572492.1 | 2 - 3 | 492 | 63 | ||
| NM_167161.2 | NP_727276.1 | 3 - 4 | 496 | 63 | ||
| GenBank mRNA | AY058283.1 | - | 2 - 3 | 492 | 63 | |
Set Membership: |
|