Search
Search

rudhira| Gene Symbol: rudhira | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258702.1 | NP_001245631.1 | 5 - 6 | 3349 | 74 | |
| NM_001258703.2 | NP_001245632.1 | 4 - 5 | 3551 | 74 | ||
| NM_001272534.1 | NP_001259463.1 | 4 - 5 | 2611 | 74 | ||
| NM_001382106.1 | NP_001368979.1 | 4 - 5 | 3551 | 74 | ||
| NM_132522.3 | NP_572750.3 | 4 - 5 | 3551 | 74 | ||
| NM_167309.3 | NP_727567.1 | 4 - 5 | 3551 | 74 | ||
| GenBank mRNA | AY075392.1 | - | 3 - 4 | 2363 | 74 | |
Set Membership: |
|