Search
Search

sbr| Gene Symbol: sbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001369981.1 | NP_001356941.1 | 4 - 5 | 694 | 83 | |
| NM_001369982.1 | NP_001356942.1 | 3 - 4 | 931 | 83 | ||
| NM_079921.3 | NP_524660.1 | 6 - 7 | 1521 | 83 | ||
| GenBank mRNA | AJ251947.2 | - | 6 - 7 | 1485 | 83 | |
| AJ318090.1 | - | 6 - 7 | 1520 | 83 | ||
Set Membership: |
|