Search
Search

AstA-R1| Gene Symbol: AstA-R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079961.4 | NP_524700.1 | 6 - 7 | 1328 | 66 | |
| NM_166982.2 | NP_726877.2 | 5 - 6 | 1279 | 66 | ||
| GenBank mRNA | AF163775.1 | - | 4 - 5 | 793 | 66 | |
| AF220216.1 | - | 6 - 7 | 1066 | 66 | ||
| BT150204.1 | - | 6 - 7 | 1331 | 66 | ||
Set Membership: |
|