Search
Search

insb| Gene Symbol: insb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299602.1 | NP_001286531.1 | 1 | 953 | 118 | |
| NM_137372.4 | NP_611216.1 | 1 | 748 | 118 | ||
| GenBank mRNA | AY071537.1 | - | 1 | 744 | 118 | |
Set Membership: |
|