Search
Search

Sh| Gene Symbol: Sh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201739.2 | NP_001188668.1 | 12 - 13 | 3889 | 100 | |
| NM_001316640.1 | NP_001303569.1 | 11 - 12 | 2142 | 100 | ||
| NM_167594.4 | NP_728122.1 | 12 - 13 | 3889 | 100 | ||
| NM_167595.4 | NP_728123.1 | 11 - 12 | 2142 | 100 | ||
| NM_206775.5 | NP_996498.2 | 14 - 15 | 3496 | 100 | ||
| GenBank mRNA | X07132.1 | - | 1770 | 100 | ||
Set Membership: |
|