Search
Search

CoRest| Gene Symbol: CoRest | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014752.4 | NP_001014752.2 | 2 - 3 | 601 | 108 | |
| NM_001014753.3 | NP_001014753.1 | 2 - 3 | 601 | 108 | ||
| NM_001014754.1 | NP_001014754.1 | 3 - 4 | 562 | 108 | ||
| NM_001014755.2 | NP_001014755.1 | 2 - 3 | 847 | 108 | ||
| NM_001014756.2 | NP_001014756.1 | 2 - 3 | 601 | 108 | ||
| NM_001201742.3 | NP_001188671.1 | 2 - 3 | 601 | 108 | ||
| NM_001258824.1 | NP_001245753.1 | 2 - 3 | 681 | 108 | ||
| NM_001258825.2 | NP_001245754.1 | 2 - 3 | 781 | 108 | ||
| NM_001272777.1 | NP_001259706.1 | 2 - 3 | 609 | 108 | ||
| GenBank mRNA | BT023817.1 | - | 2 - 3 | 579 | 108 | |
| BT099711.1 | - | 2 - 3 | 505 | 108 | ||
Set Membership: |
|