Search
Search

hydra| Gene Symbol: hydra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169341.2 | NP_001162812.2 | 3 - 4 | 743 | 83 | |
| NM_001258852.1 | NP_001245781.1 | 3 - 4 | 742 | 83 | ||
| NM_001258853.1 | NP_001245782.1 | 3 - 4 | 760 | 83 | ||
| NM_001258854.1 | NP_001245783.1 | 3 - 4 | 758 | 83 | ||
| NM_001258855.1 | NP_001245784.1 | 3 - 4 | 759 | 83 | ||
| NM_001258856.1 | NP_001245785.1 | 3 - 4 | 749 | 83 | ||
| NM_134561.7 | NP_608405.4 | 3 - 4 | 882 | 83 | ||
| NM_167711.3 | NP_728358.1 | 3 - 4 | 724 | 83 | ||
Set Membership: |
|