Search
Search

Dic61B| Gene Symbol: Dic61B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138157.3 | NP_612001.1 | 3 - 4 | 647 | 74 | |
| NM_167796.3 | NP_728477.1 | 3 - 4 | 621 | 74 | ||
| GenBank mRNA | AY069035.1 | - | 3 - 4 | 561 | 74 | |
Set Membership: |
|