Search
Search

RpS16| Gene Symbol: RpS16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299802.1 | NP_001286731.1 | 5 - 6 | 517 | 69 | |
| NM_137841.4 | NP_611685.1 | 4 - 5 | 336 | 69 | ||
| GenBank mRNA | AY070671.1 | - | 4 - 5 | 342 | 69 | |
| KX532104.1 | - | 3 - 4 | 305 | 69 | ||
Set Membership: |
|