Search
Search

Ekar| Gene Symbol: Ekar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042785.2 | NP_001036250.1 | 4 - 5 | 681 | 164 | |
| NM_001042786.2 | NP_001036251.2 | 4 - 5 | 681 | 164 | ||
| NM_143674.2 | NP_651931.1 | 4 - 5 | 681 | 164 | ||
Set Membership: |
|