Search
Search

Iml1| Gene Symbol: Iml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274330.1 | NP_001261259.1 | 2 - 3 | 490 | 75 | |
| NM_001274331.1 | NP_001261260.1 | 2 - 3 | 490 | 75 | ||
| NM_139361.2 | NP_647618.1 | 2 - 3 | 490 | 75 | ||
| NM_167888.2 | NP_728620.1 | 2 - 3 | 490 | 75 | ||
| NM_167889.2 | NP_728621.1 | 2 - 3 | 490 | 75 | ||
Set Membership: |
|