Search
Search

Ythdc1| Gene Symbol: Ythdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139554.4 | NP_647811.2 | 2 - 3 | 1055 | 66 | |
| NM_168029.2 | NP_728876.1 | 2 - 3 | 941 | 66 | ||
| GenBank mRNA | BT150336.1 | - | 2 - 3 | 1051 | 66 | |
Set Membership: |
|