Search
Search

frm| Gene Symbol: frm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144416.3 | NP_001137888.1 | 3 - 4 | 543 | 123 | |
| NM_001144417.3 | NP_001137889.1 | 3 - 4 | 543 | 123 | ||
| NM_139713.5 | NP_647970.1 | 4 - 5 | 327 | 123 | ||
| NM_168119.4 | NP_729073.1 | 3 - 4 | 543 | 123 | ||
| GenBank mRNA | AY069625.1 | - | 3 - 4 | 307 | 123 | |
Set Membership: |
|