Search
Search

Lim1| Gene Symbol: Lim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272429.2 | NP_001259358.1 | 3 - 4 | 580 | 76 | |
| NM_132277.3 | NP_572505.1 | 5 - 6 | 1674 | 76 | ||
| GenBank mRNA | AB034690.1 | - | 5 - 6 | 1645 | 76 | |
| AF181631.2 | - | 5 - 6 | 1646 | 76 | ||
Set Membership: |
|