Search
Search

DIP2| Gene Symbol: DIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259601.2 | NP_001246530.1 | 4 - 5 | 1258 | 61 | |
| NM_001274262.1 | NP_001261191.1 | 4 - 5 | 1261 | 61 | ||
| NM_001274263.1 | NP_001261192.1 | 4 - 5 | 1258 | 61 | ||
| NM_001274264.1 | NP_001261193.1 | 4 - 5 | 1261 | 61 | ||
| NM_138175.4 | NP_612019.2 | 4 - 5 | 1261 | 61 | ||
Set Membership: |
|