Search
Search

Argk| Gene Symbol: Argk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168311.3 | NP_729446.1 | 3 - 4 | 935 | 68 | |
| NM_168312.3 | NP_729447.1 | 2 - 3 | 698 | 68 | ||
| GenBank mRNA | AY069099.1 | - | 3 - 4 | 737 | 68 | |
Set Membership: |
|