Search
Search

ome| Gene Symbol: ome | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104136.2 | NP_001097606.1 | 6 - 7 | 2414 | 99 | |
| NM_001144483.2 | NP_001137955.1 | 5 - 6 | 2453 | 99 | ||
| NM_001274915.1 | NP_001261844.1 | 4 - 5 | 1607 | 99 | ||
| NM_080000.4 | NP_524739.1 | 5 - 6 | 1700 | 99 | ||
| GenBank mRNA | AY058514.1 | - | 5 - 6 | 1673 | 99 | |
| BT024224.1 | - | 5 - 6 | 2424 | 99 | ||
| BT029957.1 | - | 6 - 7 | 2398 | 99 | ||
Set Membership: |
|