Search
Search

msk| Gene Symbol: msk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080041.3 | NP_524780.1 | 1 - 2 | 348 | 55 | |
| GenBank mRNA | AF132299.1 | - | 1 - 2 | 257 | 55 | |
| BT053682.1 | - | 1 - 2 | 349 | 55 | ||
Set Membership: |
|