Search
Search

Paics| Gene Symbol: Paics | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272564.1 | NP_001259493.1 | 2 - 3 | 473 | 71 | |
| NM_132598.4 | NP_572826.1 | 2 - 3 | 476 | 71 | ||
| GenBank mRNA | AF102579.1 | - | 2 - 3 | 458 | 71 | |
| AY095520.1 | - | 2 - 3 | 441 | 71 | ||
| BT126073.1 | - | 2 - 3 | 471 | 71 | ||
Set Membership: |
|