Search
Search

rad| Gene Symbol: rad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298247.1 | NP_001285176.1 | 3 - 4 | 1036 | 117 | |
| NM_001298248.1 | NP_001285177.1 | 3 - 4 | 1260 | 117 | ||
| NM_001298249.1 | NP_001285178.1 | 3 - 4 | 1036 | 117 | ||
| NM_001298250.1 | NP_001285179.1 | 3 - 4 | 1260 | 117 | ||
| NM_001298251.1 | NP_001285180.1 | 3 - 4 | 1036 | 117 | ||
Set Membership: |
|