Search
Search

ppk23| Gene Symbol: ppk23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014749.2 | NP_001014749.1 | 3 - 4 | 847 | 89 | |
| NM_001103541.3 | NP_001097011.1 | 3 - 4 | 847 | 89 | ||
| NM_001272717.1 | NP_001259646.1 | 3 - 4 | 847 | 89 | ||
| NM_132992.3 | NP_573220.2 | 3 - 4 | 847 | 89 | ||
| GenBank mRNA | AY226550.1 | - | 3 - 4 | 754 | 89 | |
Set Membership: |
|