Search
Search

Rh50| Gene Symbol: Rh50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274505.2 | NP_001261434.1 | 2 - 3 | 664 | 81 | |
| NM_001300009.1 | NP_001286938.1 | 2 - 3 | 664 | 81 | ||
| NM_001382192.1 | NP_001369039.1 | 2 - 3 | 664 | 81 | ||
| NM_139683.3 | NP_647940.1 | 2 - 3 | 664 | 81 | ||
| GenBank mRNA | AF172639.1 | - | 2 - 3 | 646 | 81 | |
| AF181624.1 | - | 2 - 3 | 646 | 81 | ||
Set Membership: |
|