Search
Search

Bre1| Gene Symbol: Bre1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300021.1 | NP_001286950.1 | 2 - 3 | 1289 | 80 | |
| NM_139732.3 | NP_647989.2 | 2 - 3 | 1289 | 80 | ||
| GenBank mRNA | BT082047.1 | - | 2 - 3 | 1288 | 80 | |
Set Membership: |
|