Search
Search

her| Gene Symbol: her | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273577.1 | NP_001260506.1 | 1 - 2 | 1068 | 60 | |
| NM_057224.5 | NP_476572.1 | 1 - 2 | 1068 | 60 | ||
| GenBank mRNA | AY058630.1 | - | 1 - 2 | 1033 | 60 | |
Set Membership: |
|