Search
Search

Lam| Gene Symbol: Lam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258963.2 | NP_001245892.1 | 2 - 3 | 1398 | 71 | |
| NM_001258964.2 | NP_001245893.1 | 3 - 4 | 1667 | 71 | ||
| NM_001298700.1 | NP_001285629.1 | 3 - 4 | 1354 | 71 | ||
| NM_057268.5 | NP_476616.1 | 3 - 4 | 1292 | 71 | ||
| GenBank mRNA | BT001506.1 | - | 3 - 4 | 1262 | 71 | |
Set Membership: |
|