Search
Search

ush| Gene Symbol: ush | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272899.1 | NP_001259828.1 | 4 - 5 | 1909 | 65 | |
| NM_001272900.1 | NP_001259829.1 | 4 - 5 | 1709 | 65 | ||
| NM_001272901.1 | NP_001259830.1 | 4 - 5 | 1497 | 65 | ||
| NM_001272902.1 | NP_001259831.1 | 5 - 6 | 3339 | 65 | ||
| NM_057432.3 | NP_476780.1 | 4 - 5 | 1979 | 65 | ||
| GenBank mRNA | BT015278.1 | - | 4 - 5 | 1692 | 65 | |
Set Membership: |
|