Search
Search

Vinc| Gene Symbol: Vinc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297884.1 | NP_001284813.1 | 3 - 4 | 2443 | 78 | |
| NM_001297885.1 | NP_001284814.1 | 3 - 4 | 2537 | 78 | ||
| NM_057472.3 | NP_476820.1 | 3 - 4 | 2465 | 78 | ||
| GenBank mRNA | AY128501.1 | - | 3 - 4 | 2395 | 78 | |
Set Membership: |
|