Search
Search

Dcp-1| Gene Symbol: Dcp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057626.4 | NP_476974.1 | 1 - 2 | 558 | 67 | |
| GenBank mRNA | AF001464.1 | - | 1 - 2 | 508 | 67 | |
| BT010065.1 | - | 1 - 2 | 534 | 67 | ||
Set Membership: |
|