Search
Search

Cyp4e2| Gene Symbol: Cyp4e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299267.1 | NP_001286196.1 | 3 - 4 | 434 | 127 | |
| NM_057769.4 | NP_477117.2 | 2 - 3 | 337 | 127 | ||
| GenBank mRNA | AY058518.1 | - | 2 - 3 | 313 | 127 | |
Set Membership: |
|