Search
Search

vkg| Gene Symbol: vkg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273142.1 | NP_001260071.1 | 4 - 5 | 487 | 74 | |
| NM_001273143.1 | NP_001260072.1 | 4 - 5 | 562 | 74 | ||
| NM_057842.3 | NP_477190.1 | 4 - 5 | 508 | 74 | ||
Set Membership: |
|