Search
Search

kraken| Gene Symbol: kraken | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272918.1 | NP_001259847.1 | 2 - 3 | 244 | 77 | |
| NM_057917.4 | NP_477265.1 | 2 - 3 | 324 | 77 | ||
| GenBank mRNA | AY095092.1 | - | 2 - 3 | 325 | 77 | |
| AY118303.1 | - | 2 - 3 | 306 | 77 | ||
Set Membership: |
|