Search
Search

mys| Gene Symbol: mys | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298069.1 | NP_001284998.1 | 4 - 5 | 1161 | 71 | |
| NM_001298070.1 | NP_001284999.1 | 4 - 5 | 1371 | 71 | ||
| NM_080054.3 | NP_524793.2 | 4 - 5 | 1016 | 71 | ||
| GenBank mRNA | AY113499.1 | - | 4 - 5 | 1014 | 71 | |
Set Membership: |
|