Search
Search

wek| Gene Symbol: wek | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273543.1 | NP_001260472.1 | 2 - 3 | 1153 | 71 | |
| NM_080191.3 | NP_524930.1 | 2 - 3 | 1153 | 71 | ||
| GenBank mRNA | AY051673.1 | - | 2 - 3 | 1121 | 71 | |
Set Membership: |
|