Search
Search

Scgdelta| Gene Symbol: Scgdelta | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169168.2 | NP_001162639.2 | 2 - 3 | 942 | 69 | |
| NM_001169169.2 | NP_001162640.1 | 2 - 3 | 936 | 69 | ||
| NM_001169170.2 | NP_001162641.1 | 1 - 2 | 1129 | 69 | ||
| NM_080300.3 | NP_525039.1 | 2 - 3 | 942 | 69 | ||
| GenBank mRNA | AF277393.1 | - | 1 - 2 | 687 | 69 | |
| BT064640.1 | - | 2 - 3 | 934 | 69 | ||
Set Membership: |
|