Search
Search

nAChRalpha3| Gene Symbol: nAChRalpha3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298086.1 | NP_001285015.1 | 4 - 5 | 1714 | 56 | |
| NM_080340.4 | NP_525079.3 | 4 - 5 | 1714 | 56 | ||
Set Membership: |
|