Search
Search

mthl3| Gene Symbol: mthl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299597.1 | NP_001286526.1 | 6 - 7 | 1720 | 72 | |
| NM_001299598.1 | NP_001286527.1 | 6 - 7 | 1471 | 72 | ||
| NM_145335.3 | NP_663308.1 | 6 - 7 | 1409 | 72 | ||
Set Membership: |
|