Search
Search

e(y)3| Gene Symbol: e(y)3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272789.1 | NP_001259718.1 | 10 - 11 | 5381 | 66 | |
| NM_001272790.1 | NP_001259719.1 | 10 - 11 | 6216 | 66 | ||
| NM_001272791.1 | NP_001259720.1 | 10 - 11 | 6734 | 66 | ||
| NM_001272792.1 | NP_001259721.1 | 11 - 12 | 5547 | 66 | ||
| NM_134490.3 | NP_608334.3 | 10 - 11 | 5953 | 66 | ||
| GenBank mRNA | BT099689.1 | - | 10 - 11 | 5936 | 66 | |
Set Membership: |
|