Search
Search

Pmp70| Gene Symbol: Pmp70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272803.1 | NP_001259732.1 | 3 - 4 | 430 | 72 | |
| NM_001272804.1 | NP_001259733.1 | 2 - 3 | 584 | 72 | ||
| NM_001272805.1 | NP_001259734.1 | 2 - 3 | 887 | 72 | ||
| NM_134510.2 | NP_608354.1 | 3 - 4 | 677 | 72 | ||
| GenBank mRNA | AY118902.1 | - | 3 - 4 | 647 | 72 | |
Set Membership: |
|