Search
Search

CycK| Gene Symbol: CycK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299202.1 | NP_001286131.1 | 1 | 1760 | 122 | |
| NM_176068.3 | NP_788082.1 | 1 | 1760 | 122 | ||
| NM_176069.4 | NP_788083.1 | 2 | 1560 | 122 | ||
| GenBank mRNA | AY051667.1 | - | 1 | 1760 | 122 | |
Set Membership: |
|