Search
Search

Pkn| Gene Symbol: Pkn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001201966.2 | NP_001188895.1 | 3 - 4 | 981 | 57 | |
| NM_001259292.2 | NP_001246221.1 | 2 - 3 | 573 | 57 | ||
| NM_176112.2 | NP_788292.1 | 2 - 3 | 564 | 57 | ||
| NM_176113.3 | NP_788293.2 | 2 - 3 | 564 | 57 | ||
Set Membership: |
|