Search
Search

Hs3st-A| Gene Symbol: Hs3st-A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299660.1 | NP_001286589.1 | 8 - 9 | 2999 | 87 | |
| NM_176229.3 | NP_788409.1 | 8 - 9 | 2999 | 87 | ||
| GenBank mRNA | AY121626.1 | - | 8 - 9 | 2984 | 87 | |
Set Membership: |
|