Search
Search

stet| Gene Symbol: stet | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176272.2 | NP_788450.1 | 7 - 8 | 1292 | 69 | |
| NM_176273.2 | NP_788451.1 | 3 - 4 | 810 | 69 | ||
| GenBank mRNA | AY119150.1 | - | 3 - 4 | 797 | 69 | |
Set Membership: |
|